Learn More
Polyscience 15L Refrigerated Circulator, Standard Digital Controller (-30° to 170°C); 120V, 60Hz
SDP
Catalog No. 501130584
Encompass_Preferred
PolyScience™ 15 Liter Refrigerated CirculatorStandard Digital Controller-30 to 170° C Working Temperature RangeTemperature Stability: ±0.04°CCooling Capacity at 20° C: 915W1100W HeaterMaximum Pressure: 4.3 psi (0.30 bar) Maximum Pressure Flow Rate: 2.9 gpm (11 LPM) 120V, 60HzFeatures:• Large, universal icon and English display• Swivel 180™ Rotating Controller• On-board connectivity: RS-232 serial output• On-screen prompts• Automatic performance optimization• Single-point calibration capability
Catalog No. 50-113-0584 Supplier Polyscience Supplier No. SD15R30A11B
Add to Cart
Add to Cart
Request a Quote
PolyScience™ 15 Liter Refrigerated CirculatorStandard Digital Controller-30 to 170° C Working Temperature RangeTemperature Stability: ±0.04°CCooling Capacity at 20° C: 915W1100W HeaterMaximum Pressure: 4.3 psi (0.30 bar) Maximum Pressure Flow Rate: 2.9 gpm (11 LPM) 120V, 60HzFeatures:• Large, universal icon and English display• Swivel 180™ Rotating Controller• On-board connectivity: RS-232 serial output• On-screen prompts• Automatic performance optimization• Single-point calibration capability