Learn More
Polyscience 7L Space-Saving Refrigerated Circulator, Standard Digital Controller (-20 to 170°C); 120V, 60Hz
SDP
Catalog No. 501130581
Encompass_Preferred
PolyScience™ 7 Liter Space-Saving Refrigerated CirculatorStandard Digital Controller-20 to 170°C Working Temperature RangeCooling Capacity at 20°C: 200W1100W HeaterMaximum Pressure: 3.5 psi (0.24 bar) Maximum Pressure Flow Rate: 2.9 gpm (11 LPM)120V, 60HzFeatures:• Large, universal icon and English display• Swivel 180™ Rotating Controller• On-board connectivity: RS-232 serial output• On-screen prompts• Automatic performance optimization• Single-point calibration capability
Catalog No. 50-113-0581 Supplier Polyscience Supplier No. SD07R20A11B
PolyScience™ 7 Liter Space-Saving Refrigerated CirculatorStandard Digital Controller-20 to 170°C Working Temperature RangeCooling Capacity at 20°C: 200W1100W HeaterMaximum Pressure: 3.5 psi (0.24 bar) Maximum Pressure Flow Rate: 2.9 gpm (11 LPM)120V, 60HzFeatures:• Large, universal icon and English display• Swivel 180™ Rotating Controller• On-board connectivity: RS-232 serial output• On-screen prompts• Automatic performance optimization• Single-point calibration capability