Learn More
Aviva Systems Biology Corp RFK Antibody - middle region (ARP64673_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64673_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFK. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RFK. Alternate names: RIFKApplicationWB. Species reactivity: Human
Catalog No. 50-288-331