Learn More
Proteintech Group Inc RFP-Booster Alexa Fluor 568
SDP
Catalog No. 502258739 Shop All Proteintech Group Inc Products
Encompass
anti-RFP VHH/ Nanobody conjugated to fluorophore for IF, of mCherry and other red fluorescent protein-fusion proteins
This item is not returnable. View return policy
Catalog No. 50-225-8739 Supplier Proteintech Group Inc Supplier No. RB2AF56850
anti-RFP VHH/ Nanobody conjugated to fluorophore for IF, of mCherry and other red fluorescent protein-fusion proteins