Learn More
Gentarget RFP (CMV, Zeo) lentiviral particles in PBS
Catalog No. NC3364805
Encompass
Pre-made RFP lentiviral particles expressing codon optimized Red Fluorescent Protein (RFP) marker under suCMV promoter. A Zeocin antibiotic selection under Rsv promoter allows to select transduced fluorescent positive cells. The following scheme showed the lentivector expression cassette:Amount: 1 x 108 IFU/ml x 200ul in PBS. (This is the concentrated lentivirus with higher titer)
Catalog No. NC3364805 Supplier Gentarget Supplier No. LVP1224PBS
Pre-made RFP lentiviral particles expressing codon optimized Red Fluorescent Protein (RFP) marker under suCMV promoter. A Zeocin antibiotic selection under Rsv promoter allows to select transduced fluorescent positive cells. The following scheme showed the lentivector expression cassette:Amount: 1 x 108 IFU/ml x 200ul in PBS. (This is the concentrated lentivirus with higher titer)