Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Red fluorescent protein, Discosoma sp. (His-tag) | >85.0% | 50UG
SDP

Catalog No. 5000176012
Encompass_Preferred

Recombinant red fluorescent protein derived from Discosoma sp., expressed in Escherichia coli and supplied as a lyophilized powder with a C-terminal 6xHis affinity tag. The protein corresponds to a 225-amino-acid red fluorescent protein and is provided for research use with >85% purity; store at -20°C and reconstitute to at least 100 μg/mL in ddH2O.

  • Provides red fluorescence for labeling and imaging.
  • Expressed in Escherichia coli for scalable production.
  • Includes a C-terminal 6xHis tag for affinity purification.
  • Supplied as lyophilized powder for long-term storage.
  • Purity greater than 85% by reducing SDS-PAGE.
  • Recommended reconstitution concentration of ≥100 μg/mL.
  • Store at -20°C; stable at 4°C for 1 week after reconstitution.
  • Intended for research use only.

Catalog No. 50-001-76012 Supplier Medchemexpress LLC Supplier No. HYP70306850UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant red fluorescent protein derived from Discosoma sp., expressed in Escherichia coli and supplied as a lyophilized powder with a C-terminal 6xHis affinity tag. The protein corresponds to a 225-amino-acid red fluorescent protein and is provided for research use with >85% purity; store at -20°C and reconstitute to at least 100 μg/mL in ddH2O.

  • Provides red fluorescence for labeling and imaging.
  • Expressed in Escherichia coli for scalable production.
  • Includes a C-terminal 6xHis tag for affinity purification.
  • Supplied as lyophilized powder for long-term storage.
  • Purity greater than 85% by reducing SDS-PAGE.
  • Recommended reconstitution concentration of ≥100 μg/mL.
  • Store at -20°C; stable at 4°C for 1 week after reconstitution.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.