Learn More
Angio Proteomie RFP Expressing Human Umbilical Vein Endothelial Cells (RFP-HUVECs), Red Fluorescent Protein (RFP), 5 x 10^5 cells/vial, Passage 3, Zeocin resistant, >18 minimum population doublings
SDP
Catalog No. NC0731622
Encompass
RFP Expressing Human Umbilical Vein Endothelial Cells (RFP-HUVECs), Red Fluorescent Protein (RFP), 5 x 10^5 cells/vial, Passage 3, Zeocin resistant, >18 minimum population doublings
Catalog No. NC0731622 Supplier Angio Proteomie Supplier No. CAP0001RFP
RFP Expressing Human Umbilical Vein Endothelial Cells (RFP-HUVECs), Red Fluorescent Protein (RFP), 5 x 10^5 cells/vial, Passage 3, Zeocin resistant, >18 minimum population doublings