Learn More
Proteintech Group Inc RFP-Trap Agarose
SDP
Supplier:  Proteintech Group Inc RTA10
Encompass
RFP-Trap Agarose is an affinity resin for IP of RFP-fusion proteins. It consists of an RFP Nanobody/ VHH coupled to agarose beads.
Catalog No. 50-225-8746