Learn More
Proteintech Group Inc RFP-Trap Magnetic Agarose
SDP
Supplier:  Proteintech Group Inc RTMA20
Encompass
The ChromoTek RFP-Trap Magnetic Agarose are affinity beads for IP of RFP-fusion proteins. It comprises a RFP Nanobody/ VHH coupled to magnetic agarose.
Catalog No. 50-225-8761