Learn More
Aviva Systems Biology Corp RFP2 Antibody - middle region (ARP34144_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34144_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RFP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RFP2. Alternate names: CAR, LEU5, RFP2, DLEU5, RNF77ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55270