Learn More
Aviva Systems Biology Corp RFP2 Antibody - middle region (ARP34143_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34143_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFP2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human RFP2. Alternate names: CAR, LEU5, RFP2, DLEU5, RNF77ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55269