Learn More
Aviva Systems Biology Corp RFPL3 Antibody - C-terminal region (ARP43077_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43077_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFPL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RFPL3. Alternate names: RP1-149A16.1ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-58418