Learn More
Bachem RFRP-3 (rat)
SDP
Supplier:  Bachem 4040360.0005
Encompass_Preferred
RFRP-3 (rat) , H-Ala-Asn-Met-Glu-Ala-Gly-Thr-Met-Ser-His-Phe-Pro-Ser-Leu-Pro-Gln-Arg-Phe-NH2, 420088-80-8, C88H134N26O25S2, Mr 2020.32, 5mg. Neuropeptide VF Precursor (108-125) amide (rat), RFamide-Related Peptide 3 (rat), NPVF (rat). RFRP-3 belongs to a family of RFamide peptides termed RFamide-related peptides (RFRPs), reported to be present in the mammalian brain. The preproprotein of rat RFRPs contains two putative peptides RFRP-1 and RFRP-3 and it could be demonstrated by K.Ukena et al., that the octadecapeptide ANMEAGTMSHFPSLPQRF represents the mature form of RFPR-3 in the rat hypothalamus.
Catalog No. 50-194-7473