Learn More
Aviva Systems Biology Corp RFTN2 Antibody - N-terminal region (ARP52707_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52707_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFTN2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human RFTN2. Alternate names: C2orf11, FLJ30574, MGC117313, Raftlin-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-758