Learn More
Aviva Systems Biology Corp COP1 Antibody - middle region (ARP42955_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42955_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rfwd2. It was validated on Western Blot. . Alternate names: AI316802, C80879, Cop1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-048