Learn More
Aviva Systems Biology Corp RFX1 Antibody - N-terminal region (ARP38199_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38199_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RFX1. Alternate names: EF-C, EFC, RFXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56560