Learn More
Aviva Systems Biology Corp RFX2 Antibody - middle region (ARP38004_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38004_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RFX2. Alternate names: FLJ14226ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56483