Learn More
Aviva Systems Biology Corp RFX4 Antibody - N-terminal region (ARP40086_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40086_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFX4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RFX4. Alternate names: NYD-SP10ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-57290