Learn More
Aviva Systems Biology Corp RFX4 Antibody - N-terminal region (ARP39699_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39699_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RFX4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RFX4. Alternate names: NYD-SP10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57142