Learn More
Aviva Systems Biology Corp RFX5 Antibody - N-terminal region (ARP37992_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37992_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RFX5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RFX5. Alternate names: ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-56478