Learn More
Aviva Systems Biology Corp Rfxap Antibody - middle region (ARP37454_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37454_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rfxap. It was validated on Western Blot. Rfxap,regulatory factor X-associated protein,5730495K23Rik; Rfxap,regulatory factor X-associated protein,regulatory factor X-associated protein,RFX-associated proteinThe immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 5730495K23RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-636