Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RG-101 sodium | 00-00-0 | 95.4% | 8537.6 (free acid) | C295H410N78Na19O146P19S17 | 5 MG
SDP

Catalog No. 5000263256
Encompass_Preferred

RG-101 sodium is a hepatocyte-targeted N-acetylgalactosamine (GalNAc)-conjugated antisense oligonucleotide that antagonizes microRNA-122 (miR-122), a host factor important for hepatitis C virus replication. Supplied as a sodium salt, it is used in research to study miR-122 biology and antiviral mechanisms.

  • Hepatocyte-targeted GalNAc conjugate for liver delivery.
  • Antagonizes miR-122 to modulate hepatitis C virus-related pathways.
  • High molecular weight oligonucleotide for sequence-specific activity.
  • High purity (95.4%) suitable for research applications.
  • Soluble in water; sonication may be required for dissolution.
  • Recommended storage at -20°C; in solution store at -80°C for long-term.

Catalog No. 50-002-63256 Supplier Medchemexpress LLC Supplier No. HY132600A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RG-101 sodium is a hepatocyte-targeted N-acetylgalactosamine (GalNAc)-conjugated antisense oligonucleotide that antagonizes microRNA-122 (miR-122), a host factor important for hepatitis C virus replication. Supplied as a sodium salt, it is used in research to study miR-122 biology and antiviral mechanisms.

  • Hepatocyte-targeted GalNAc conjugate for liver delivery.
  • Antagonizes miR-122 to modulate hepatitis C virus-related pathways.
  • High molecular weight oligonucleotide for sequence-specific activity.
  • High purity (95.4%) suitable for research applications.
  • Soluble in water; sonication may be required for dissolution.
  • Recommended storage at -20°C; in solution store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.