Learn More
Apexbio Technology LLC RG 108, 10mg. Cas: 48208-26-0 MFCD: MFCD08705332. DNA methyltransferase inhibitor
SDP
Supplier:  Apexbio Technology LLC A191310
Encompass_Preferred
RG108 is a DNA methyltransferase (DMNT) inhibitor that enhanced reprogramming of OK-transduced MEFs in the presence of BIX. It is a small molecule that effectively blocked DNA methyltransferases in vitro and did not cause covalent enzyme trapping in human cell lines. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4722