Learn More
Abcam RG 108, non-nucleoside DNA methyltransferase inhibitor (DNMT), 10MG
Catalog No. 502706972
Encompass_Preferred
MW 334.3 Da, Purity >99%. Novel, non-nucleoside DNA methyltransferase (DNMT) inhibitor, capable of interacting with the DNMT1 active site (IC₅₀ = 115 nM). Inhibits DNA methylation in human cancer cell lines. Reactivates several epigenetically silenced tumour suppressor genes in vitro. Capable of enhancing the efficiency of induced pluripotent stem cell (iPS) generation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6972 Supplier Abcam Supplier No. AB14101310MG
MW 334.3 Da, Purity >99%. Novel, non-nucleoside DNA methyltransferase (DNMT) inhibitor, capable of interacting with the DNMT1 active site (IC₅₀ = 115 nM). Inhibits DNA methylation in human cancer cell lines. Reactivates several epigenetically silenced tumour suppressor genes in vitro. Capable of enhancing the efficiency of induced pluripotent stem cell (iPS) generation.
The product is subject to the following: Abcam Restricted Use Statement