Learn More
Medchemexpress LLC Rg-12525 | 120128-20-3 | 98.3% | 423.47 g/mol | C25H21N5O2 | 25 MG
SDP
Catalog No. 5000248597
Encompass_Preferred
RG-12525 is a small-molecule research compound reported as a potent antagonist of peptidoleukotrienes (LTC4, LTD4, LTE4); it also shows PPAR-gamma agonist activity and inhibits CYP3A4. Intended for in vitro and preclinical studies, the compound is supplied at high purity and is typically dissolved in DMSO for assays.
  • Specific antagonist of peptidoleukotrienes with low-nanomolar activity
  • Shows PPAR-gamma agonist activity (approx. IC50 60 nM)
  • Inhibits CYP3A4 (Ki ≈ 0.5 μM)
  • High purity suitable for preclinical research
  • Soluble in DMSO for assay preparation
Catalog No. 50-002-48597 Supplier Medchemexpress LLC Supplier No. HY10167625MG
RG-12525 is a small-molecule research compound reported as a potent antagonist of peptidoleukotrienes (LTC4, LTD4, LTE4); it also shows PPAR-gamma agonist activity and inhibits CYP3A4. Intended for in vitro and preclinical studies, the compound is supplied at high purity and is typically dissolved in DMSO for assays.
  • Specific antagonist of peptidoleukotrienes with low-nanomolar activity
  • Shows PPAR-gamma agonist activity (approx. IC50 60 nM)
  • Inhibits CYP3A4 (Ki ≈ 0.5 μM)
  • High purity suitable for preclinical research
  • Soluble in DMSO for assay preparation