Learn More
Medchemexpress LLC Rg-12525 | 120128-20-3 | 98.3% | 423.47 g/mol | C25H21N5O2 | 50 MG
SDP
Catalog No. 5000248629
Encompass_Preferred
RG-12525 (NID 525) is a small-molecule research compound that antagonizes peptidoleukotriene receptors (LTC4, LTD4, LTE4), exhibits PPAR-γ agonist activity, and inhibits cytochrome P450 enzymes. It is provided in high-purity solid form and as a DMSO solution for biochemical and pharmacological research, supporting both in vitro and in vivo investigations.
  • Potent leukotriene receptor antagonist with low-nanomolar activity.
  • Shows PPAR-γ agonist activity (≈60 nM).
  • Inhibits CYP3A4 (Ki ≈0.5 μM), useful for metabolism studies.
  • Available as solid and DMSO solution for flexible dosing.
  • High purity suitable for biochemical and pharmacological assays.
  • Applicable to in vitro and in vivo pharmacology research.
Catalog No. 50-002-48629 Supplier Medchemexpress LLC Supplier No. HY10167650MG
RG-12525 (NID 525) is a small-molecule research compound that antagonizes peptidoleukotriene receptors (LTC4, LTD4, LTE4), exhibits PPAR-γ agonist activity, and inhibits cytochrome P450 enzymes. It is provided in high-purity solid form and as a DMSO solution for biochemical and pharmacological research, supporting both in vitro and in vivo investigations.
  • Potent leukotriene receptor antagonist with low-nanomolar activity.
  • Shows PPAR-γ agonist activity (≈60 nM).
  • Inhibits CYP3A4 (Ki ≈0.5 μM), useful for metabolism studies.
  • Available as solid and DMSO solution for flexible dosing.
  • High purity suitable for biochemical and pharmacological assays.
  • Applicable to in vitro and in vivo pharmacology research.