Learn More
Medchemexpress LLC RG-12525 | 120128-20-3 | MFCD00877828 | 98.3% | 423.47 g/mol | C25H21N5O2 | 1 ML
SDP
Catalog No. 5000245668
Encompass_Preferred
RG-12525 is a small-molecule leukotriene receptor antagonist supplied as a ready-to-use 10 mM solution in DMSO for research applications. It functions as a specific, competitive antagonist of peptidoleukotrienes (LTC4, LTD4, LTE4) and is commonly used in pharmacology and signaling studies. The compound has CAS 120128-20-3, molecular formula C25H21N5O2, molecular weight 423.47 g/mol, and reported purity of 98.3%.
  • Provided as a 10 mM solution in DMSO for ready-to-use assays.
  • Specific, competitive antagonist of peptidoleukotrienes (LTC4, LTD4, LTE4).
  • High reported purity suitable for biochemical studies.
  • Well characterized molecular properties for experimental reproducibility.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-45668 Supplier Medchemexpress LLC Supplier No. HY10167610MM 1ML
RG-12525 is a small-molecule leukotriene receptor antagonist supplied as a ready-to-use 10 mM solution in DMSO for research applications. It functions as a specific, competitive antagonist of peptidoleukotrienes (LTC4, LTD4, LTE4) and is commonly used in pharmacology and signaling studies. The compound has CAS 120128-20-3, molecular formula C25H21N5O2, molecular weight 423.47 g/mol, and reported purity of 98.3%.
  • Provided as a 10 mM solution in DMSO for ready-to-use assays.
  • Specific, competitive antagonist of peptidoleukotrienes (LTC4, LTD4, LTE4).
  • High reported purity suitable for biochemical studies.
  • Well characterized molecular properties for experimental reproducibility.
  • Intended for research use only; not for human or diagnostic use.