Learn More
Medchemexpress LLC RG-12525 | 120128-20-3 | MFCD00877828 | 98.3% | 423.47 g/mol | C25H21N5O2 | 100 MG
SDP
Catalog No. 5000246526
Encompass_Preferred
RG-12525 is a small-molecule leukotriene receptor antagonist (CAS 120128-20-3) used in pharmacology research. It antagonizes LTC4, LTD4, and LTE4, shows PPAR-gamma agonism, and inhibits CYP3A4. The compound is supplied with a reported high purity and defined solubility and storage conditions for reproducible in vitro and in vivo experiments.
  • Specific antagonist of LTC4, LTD4, and LTE4.
  • Additional PPAR-gamma agonist and CYP3A4 inhibitor activities.
  • High purity suitable for research applications (98.31%).
  • Soluble in DMSO at 25 mg/mL (requires ultrasonic).
  • Defined storage guidelines for powder and solutions to maintain stability.
Catalog No. 50-002-46526 Supplier Medchemexpress LLC Supplier No. HY101676100MG
RG-12525 is a small-molecule leukotriene receptor antagonist (CAS 120128-20-3) used in pharmacology research. It antagonizes LTC4, LTD4, and LTE4, shows PPAR-gamma agonism, and inhibits CYP3A4. The compound is supplied with a reported high purity and defined solubility and storage conditions for reproducible in vitro and in vivo experiments.
  • Specific antagonist of LTC4, LTD4, and LTE4.
  • Additional PPAR-gamma agonist and CYP3A4 inhibitor activities.
  • High purity suitable for research applications (98.31%).
  • Soluble in DMSO at 25 mg/mL (requires ultrasonic).
  • Defined storage guidelines for powder and solutions to maintain stability.