Learn More
Selleck Chemical LLC RG 13022 S2115-5mg
SDP
Catalog No. 503110383
Encompass_Preferred
RG 13022 (Tyrphostin RG13022) inhibits the autophosphorylation reaction of the EGF receptor in immunoprecipitates with IC50 of 4 M
Catalog No. 50-311-0383 Supplier Selleck Chemical LLC Supplier No. S21155MG
Add to Cart
Add to Cart
RG 13022 (Tyrphostin RG13022) inhibits the autophosphorylation reaction of the EGF receptor in immunoprecipitates with IC50 of 4 M