Learn More
Medchemexpress LLC RG-6125 1mg | 1mg
SDP
Catalog No. 5000324574
Encompass_Preferred
RG-6125 is a humanized antibody expressed in CHO targeting CDH11/Cadherin-11 RG-6125 has a huIgG2 type heavy chain and a hu type light chain with a predicted molecular weight (MW) of 145 kDa The isotype control for RG-6125 can refer to Human IgG2 kappa Isotype Control (HY-P99002)
Catalog No. 50-003-24574 Supplier Medchemexpress LLC Supplier No. HYP9906561MG
RG-6125 is a humanized antibody expressed in CHO targeting CDH11/Cadherin-11 RG-6125 has a huIgG2 type heavy chain and a hu type light chain with a predicted molecular weight (MW) of 145 kDa The isotype control for RG-6125 can refer to Human IgG2 kappa Isotype Control (HY-P99002)