Learn More
Selleck Chemical LLC RG-7112
SDP
Catalog No. 5000002436
Encompass_Preferred
RG7112 (RO5045337) is an orally bioavailable and selective p53-MDM2 inhibitor with HTRF IC50 of 18 nM
Catalog No. 50-000-02436 Supplier Selleck Chemical LLC Supplier No. S7030100MG
Add to Cart
Add to Cart
RG7112 (RO5045337) is an orally bioavailable and selective p53-MDM2 inhibitor with HTRF IC50 of 18 nM