Learn More
Selleck Chemical LLC RG-7112 5mg 939981-39-2 RO5045337
SDP
Catalog No. 501364334
Encompass_Preferred
RG7112 (RO5045337) is an orally bioavailable and selective p53-MDM2 inhibitor with HTRF IC50 of 18 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4334 Supplier Selleck Chemical LLC Supplier No. S70305MG
RG7112 (RO5045337) is an orally bioavailable and selective p53-MDM2 inhibitor with HTRF IC50 of 18 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.