Learn More
Medchemexpress LLC RG-7212 RO5458640 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9906571MG
Encompass_Preferred
RG-7212 (RO5458640) is a human-derived antibody expressed in CHO cells targeting TNFSF12/TWEAK RG-7212 (RO5458640) has a huIgG1 type heavy chain and a hu type light chain with a predicted molecular weight (MW) of 146 02 kDa The isotype control for RG-7212 (RO5458640) can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-25300