Learn More
Medchemexpress LLC RG-7652 1mg | 1mg
SDP
Catalog No. 5000325827
Encompass_Preferred
RG-7652 is a humanized antibody expressed in CHO cells targeting PCSK9 RG-7652 contains huIgG1 heavy chain and hu light chain with a predicted molecular weight (MW) of 145 88 kDa The isotype control for RG-7652 can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-25827 Supplier Medchemexpress LLC Supplier No. HYP9906581MG
RG-7652 is a humanized antibody expressed in CHO cells targeting PCSK9 RG-7652 contains huIgG1 heavy chain and hu light chain with a predicted molecular weight (MW) of 145 88 kDa The isotype control for RG-7652 can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)