Learn More
Selleck Chemical LLC RG108-10mg
SDP
Catalog No. 502390007
Encompass_Preferred
RG108 (N-Phthalyl-L-tryptophan) is an inhibitor of DNA methyltransferase with IC50 of 115 nM in a cell-free assay, does not cause trapping of covalent enzymes.
Catalog No. 50-239-0007 Supplier Selleck Chemical LLC Supplier No. S282110MG
RG108 (N-Phthalyl-L-tryptophan) is an inhibitor of DNA methyltransferase with IC50 of 115 nM in a cell-free assay, does not cause trapping of covalent enzymes.