Learn More
TARGETMOL CHEMICALS INC RG108 25MG
SDP
Catalog No. 502577240
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RG108 (N-Phthalyl-L-tryptophan) is an DNA methyltransferase inhibitor(IC50=115 nM). purity: 99%
Catalog No. 50-257-7240 Supplier TARGETMOL CHEMICALS INC Supplier No. T203825MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RG108 (N-Phthalyl-L-tryptophan) is an DNA methyltransferase inhibitor(IC50=115 nM). purity: 99%