Learn More
Selleck Chemical LLC RG108 S2821-200mg
SDP
Catalog No. 503109026
Encompass_Preferred
RG108 (N-Phthalyl-L-tryptophan) is an inhibitor of DNA methyltransferase with IC50 of 115 nM in a cell-free assay does not cause trapping of covalent enzymes
Catalog No. 50-310-9026 Supplier Selleck Chemical LLC Supplier No. S2821200MG
RG108 (N-Phthalyl-L-tryptophan) is an inhibitor of DNA methyltransferase with IC50 of 115 nM in a cell-free assay does not cause trapping of covalent enzymes