Learn More
Medchemexpress LLC HY-101426 5mg Medchemexpress, RG14620 CAS:136831-49-7 Purity:>98%
SDP
Catalog No. 501932123
Encompass_Preferred
Medchemexpress, HY-101426 5mg RG14620 CAS:136831-49-7 RG14620 is an EGFR inhibitor with an IC 50 of 3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2123 Supplier Medchemexpress LLC Supplier No. HY1014265MG
Medchemexpress, HY-101426 5mg RG14620 CAS:136831-49-7 RG14620 is an EGFR inhibitor with an IC 50 of 3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.