Learn More
Medchemexpress LLC RG2833 | 1215493-56-3 | 99.26% | 339.43 | 500 MG
SDP
Catalog No. 5000394454
Encompass_Preferred
RG2833 is a brain-penetrant HDAC inhibitor with IC50s of 60 nM and 50 nM for HDAC1 and HDAC3, respectively. The Ki values for HDAC1 and HDAC3 are 32 nM and 5 nM, respectively. It is for research use only.
  • Brain-penetrant HDAC inhibitor.
  • IC50s: 60 nM for HDAC1, 50 nM for HDAC3.
  • Ki values: 32 nM for HDAC1, 5 nM for HDAC3.
  • Highly active in the tested concentration range from 1 to 10 μM.
  • Induces sustained frataxin upregulation in cells from Friedreich's ataxia patients and in a mouse model.
  • Produces significant increases in brain aconitase enzyme activity and reduces neuronal pathology of the dorsal root ganglia (DRG).
  • Corrects frataxin deficiency in the brain and heart of KIKI mice.
  • Improves motor coordination of YG8R FRDA mice and increases frataxin protein expression in their brain.
  • Attenuates L-DOPA-induced dyskinesia in MPTP-lesioned marmosets.
Catalog No. 50-003-94454 Supplier Medchemexpress LLC Supplier No. HY16425500MG
RG2833 is a brain-penetrant HDAC inhibitor with IC50s of 60 nM and 50 nM for HDAC1 and HDAC3, respectively. The Ki values for HDAC1 and HDAC3 are 32 nM and 5 nM, respectively. It is for research use only.
  • Brain-penetrant HDAC inhibitor.
  • IC50s: 60 nM for HDAC1, 50 nM for HDAC3.
  • Ki values: 32 nM for HDAC1, 5 nM for HDAC3.
  • Highly active in the tested concentration range from 1 to 10 μM.
  • Induces sustained frataxin upregulation in cells from Friedreich's ataxia patients and in a mouse model.
  • Produces significant increases in brain aconitase enzyme activity and reduces neuronal pathology of the dorsal root ganglia (DRG).
  • Corrects frataxin deficiency in the brain and heart of KIKI mice.
  • Improves motor coordination of YG8R FRDA mice and increases frataxin protein expression in their brain.
  • Attenuates L-DOPA-induced dyskinesia in MPTP-lesioned marmosets.