Learn More
Apexbio Technology LLC RG2833, 50mg. Cas: 1215493-56-3 MFCD: MFCD25976845. Brain-penetrant HDAC inhibitor
SDP
Supplier:  Apexbio Technology LLC A376150
Encompass_Preferred
RG2833 is a histone deacetylase (HDAC) inhibitor. In the neuronal cell, RG2833 increased Friedreich Ataxia (FXN) mRNA and protein levels, with concomitant changes in the epigenetic state of the gene. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4836