Learn More
Medchemexpress LLC HY-16425 5mg Medchemexpress, RG2833 CAS:1215493-56-3 Purity:>98%
SDP
Catalog No. 502030714
Encompass_Preferred
Medchemexpress, HY-16425 5mg RG2833 CAS:1215493-56-3 RG2833 is a brain-penetrant HDAC inhibitor with IC50s of 60 nM and 50 nM for HDAC1 and HDAC3, respectively. The Ki values for HDAC1 and HDAC3 are 32 and 5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0714 Supplier Medchemexpress LLC Supplier No. HY164255MG
Medchemexpress, HY-16425 5mg RG2833 CAS:1215493-56-3 RG2833 is a brain-penetrant HDAC inhibitor with IC50s of 60 nM and 50 nM for HDAC1 and HDAC3, respectively. The Ki values for HDAC1 and HDAC3 are 32 and 5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.