Learn More
Selleck Chemical LLC RG2833 (RGFP109) 10mg 1215493-56-3
SDP
Supplier:  Selleck Chemical LLC S729210MG
Encompass_Preferred
RG2833 (RGFP109) is a brain-penetrant HDAC inhibitor with IC50 of 60 nM and 50 nM for HDAC1 and HDAC3 in cell-free assays, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4604