Learn More
Medchemexpress LLC HY-15676 100mg , RG7388 Idasanutlin CAS:1229705-06-9 Purity:98%
SDP
Catalog No. 501492751
Encompass_Preferred
Medchemexpress, HY-15676 100mg RG7388 Idasanutlin CAS:1229705-06-9 Formula:C31H29Cl2F2N3O4 IC50: 6 nM (p53-MDM2) Purity:98% RG7388 is a potent and selective MDM2 antagonist, inhibiting p53-MDM2 binding, with IC 50 of 6 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2751 Supplier Medchemexpress LLC Supplier No. HY15676100MG
Medchemexpress, HY-15676 100mg RG7388 Idasanutlin CAS:1229705-06-9 Formula:C31H29Cl2F2N3O4 IC50: 6 nM (p53-MDM2) Purity:98% RG7388 is a potent and selective MDM2 antagonist, inhibiting p53-MDM2 binding, with IC 50 of 6 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.