Learn More
Medchemexpress LLC HY-12981 5mg Medchemexpress, RG7713 CAS:920022-47-5 Purity:>98%
SDP
Catalog No. 502081221
Encompass_Preferred
Medchemexpress, HY-12981 5mg RG7713 CAS:920022-47-5 RG7713 (RO5028442) is a highly potent and selective Brain-Penetrant Vasopressin 1a (V1a) receptor antagonist with Kis of 1 nM (hV1a) and 39 nM (mV1a). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1221 Supplier Medchemexpress LLC Supplier No. HY129815MG
Medchemexpress, HY-12981 5mg RG7713 CAS:920022-47-5 RG7713 (RO5028442) is a highly potent and selective Brain-Penetrant Vasopressin 1a (V1a) receptor antagonist with Kis of 1 nM (hV1a) and 39 nM (mV1a). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.