Learn More
Medchemexpress LLC HY-101792 10mg Medchemexpress, RG7800 CAS:1449598-06-4 Purity:>98%
SDP
Catalog No. 502028680
Encompass_Preferred
Medchemexpress, HY-101792 10mg RG7800 CAS:1449598-06-4 RG7800 is a SMN2 splicing modifier. RG7800 has the potential for spinal muscular atrophy treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8680 Supplier Medchemexpress LLC Supplier No. HY10179210MG
Medchemexpress, HY-101792 10mg RG7800 CAS:1449598-06-4 RG7800 is a SMN2 splicing modifier. RG7800 has the potential for spinal muscular atrophy treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.