Learn More
Medchemexpress LLC HY-101792A 10mg , RG7800 hydrochloride CAS: Purity:>98%
SDP
Catalog No. 502260412
Encompass_Preferred
HY-101792A 10mg RG7800 hydrochloride CAS: RG7800 hydrochloride is an orally active SMN2 splicing modulator, with EC1.5xs of 23 nM and 87 nM for SMN2 splicing and SMN protein; RG7800 hydrochloride has the potential to treat spinal muscular atrophy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0412 Supplier Medchemexpress LLC Supplier No. HY101792A10MG
HY-101792A 10mg RG7800 hydrochloride CAS: RG7800 hydrochloride is an orally active SMN2 splicing modulator, with EC1.5xs of 23 nM and 87 nM for SMN2 splicing and SMN protein; RG7800 hydrochloride has the potential to treat spinal muscular atrophy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.