Learn More
Medchemexpress LLC HY-117650A 10mg Medchemexpress, RG7834 CAS:2072057-17-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY117650A10MG
Encompass_Preferred
Medchemexpress, HY-117650A 10mg RG7834 CAS:2072057-17-9 RG7834 (RO 7020322) is a highly selective and orally bioavailable HBV inhibitor, potently inhibits HBV antigens (both HBsAg and HBeAg) and HBV DNA, with IC50s of 2.8, 2.6, and 3.2 nM, respectively, in dHepaRG Cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9486