Learn More
Aviva Systems Biology Corp RG9MTD1 Antibody - middle region (ARP40877_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40877_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RG9MTD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RG9MTD1. Alternate names: FLJ20432, MRPP1, RG9MTD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-901