Learn More
Aviva Systems Biology Corp RG9MTD3 Antibody - C-terminal region (ARP41132_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41132_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RG9MTD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human RG9MTD3. Alternate names: FLJ31455, RP11-3J10.9, bA3J10.9, RG9MTD3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53278