Learn More
Medchemexpress LLC RGB-286638 CDK INHB 5MG
SDP
Catalog No. NC1698561
Encompass_Preferred
RGB-286638 CDK Inhibitor 5mg; Cas No.784210-87-3; RGB-286638 is a CDK inhibitor that inhibits the kinase activity of cyclin T1-CDK9, cyclin B1-CDK1, cyclin E-CDK2, cyclin D1-CDK4, cyclin E-CDK3, and p35-CDK5 with IC50s of 1, 2, 3, 4, 5 and 5 nM, respectively; also inhibits GSK-3β, TAK1, Jak2 and MEK1, with IC50s of 3, 5, 50, and 54 nM.
Catalog No. NC1698561 Supplier Medchemexpress LLC Supplier No. HY155045MG
RGB-286638 CDK Inhibitor 5mg; Cas No.784210-87-3; RGB-286638 is a CDK inhibitor that inhibits the kinase activity of cyclin T1-CDK9, cyclin B1-CDK1, cyclin E-CDK2, cyclin D1-CDK4, cyclin E-CDK3, and p35-CDK5 with IC50s of 1, 2, 3, 4, 5 and 5 nM, respectively; also inhibits GSK-3β, TAK1, Jak2 and MEK1, with IC50s of 3, 5, 50, and 54 nM.